3-Aminopropionitrile fumarate
Catalog # 8.41773.0025
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:256.26 g/mol
EINECS:218-208-7
CAS Number:2079-89-2
Formula:C₁₀H₁₆N₄O₄
MDL Number:MFCD00013142