Pesticide reference standard, Tolylfluanid 100 µg/ml in cyclohexane, Dr Ehrenstorfer
EHERXA17600000CY
:
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:731-27-1
- Description:Tolylfluanid 100 µg/ml in cyclohexane
- Formula:C₁₀H₁₃Cl₂FN₂O₂S₂
- Description:Tolylfluanid