Isolate DNA from soil and water samples using magnetic beads.
- Ceramic beads pre-aliquoted in a convenient 96-well format
- Unique inhibitor removal reagent
- Automation friendly
The Mag-Bind® Environmental DNA 96 Kit allows rapid and reliable isolation of high-quality DNA from soil and water samples. The Mag-Bind® Environmental DNA 96 Kit can isolate microbial DNA from yeast, fungi, and gram-positive or negative bacteria. Up to 96 one hundred mg soil samples can be processed in 120 minutes using automated liquid handlers or magnetic processors. Omega Bio-tek’s unique cHTR Reagent effectively removes humic acid and other PCR inhibitors allowing for purified DNA to be suitable for PCR, 16S sequencing, Whole Genome Sequencing, and Next-Generation Sequencing. There are no organic extractions, which reduces plastic waste and hands-on time to allow multiple samples to be processed in parallel.
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Total DNA
- Sample type:WastewaterSoil
- Target molecule:DNA