Order Entry
France
ContactUsLinkComponent
Disposable columns prepacked with Sepharose™, His SpinTrap™ kit
Disposable columns prepacked with Sepharose™, His SpinTrap™ kit
Catalog # 28-9321-71
Supplier:  Cytiva
undefined
Disposable columns prepacked with Sepharose™, His SpinTrap™ kit
Catalog # 28-9321-71
Supplier:  Cytiva
Specifications
  • Description:
    His SpinTrap™ kit
  • Binding capacity:
    At least 40 mg histidine-tagged protein/ml medium
  • Pk:
    1 KIT
Specifications
About this item
His SpinTrap™ is a prepacked, single use spin column for purifying histidine-tagged proteins by immobilised metal ion affinity chromatography (IMAC) and rapid expression screening.
  • Ni Sepharose™ High Performance in prepacked spin columns
  • Yields up to 750 µg pure protein per column
  • Allows direct purification of both clarified and unclarified cell lysates
Delivery: Kit contains one pack of His SpinTrap™ and one pack of His buffer kit.