Order Entry
France
ContactUsLinkComponent
Affinity chromatography columns, rProtein A Protein G GraviTrap™
Affinity chromatography columns, rProtein A Protein G GraviTrap™
Catalog # 28-9852-55
Supplier:  Cytiva
undefined
Affinity chromatography columns, rProtein A Protein G GraviTrap™
Catalog # 28-9852-55
Supplier:  Cytiva
Specifications
  • Capacity:
    1 ml
  • Description:
    Protein G GraviTrap™
  • Seq:
    01
  • Binding capacity:
    Approx. 20 mg human IgG/column
  • Column hardware:
    PP
  • Pk:
    10
Specifications
About this item
Protein G GraviTrap includes ten columns with 1 ml of resin for general purpose purification of polyclonal and monoclonal antibodies from most species.
  • Broad binding specificity – binds a broad range of IgG species and subclasses from most species including rat
  • High binding capacity – approximately 20 mg human IgG/ml resin
  • Simple – GraviTrap prepacked gravity flow columns enable simple one-step purification of proteins in 30 min without the need for equipment
Protein G GraviTrap columns are prepacked with Protein G Sepharose 4 Fast Flow resin. These gravity flow columns are designed for fast and efficient manual purification of monoclonal and polyclonal antibodies. Each of these protein G columns is reusable up to five times. Prepacked gravity flow columns provide convenient, fast, and reliable results.