Order Entry
France
ContactUsLinkComponent
Anion exchange chromatography columns, HiTrap™ Q XL
Anion exchange chromatography columns, HiTrap™ Q XL
Supplier:  Cytiva
Certificates
undefined
Anion exchange chromatography columns, HiTrap™ Q XL
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 17-5158-01
  • 17-5159-01
  • Max. pressure:
  • 5 bar
  • 5 bar
  • pH range:
  • 2 - 12
  • 2 - 12