Order Entry
France
ContactUsLinkComponent
Ion exchange chromatography column selection kit, HiTrap™ Capto™
Ion exchange chromatography column selection kit, HiTrap™ Capto™
Catalog # 28-9343-88
Supplier:  Cytiva
undefined
Ion exchange chromatography column selection kit, HiTrap™ Capto™
Catalog # 28-9343-88
Supplier:  Cytiva

Specifications

  • Flow rate:
    <4 ml/min
  • Ø int.:
    7,00 mm
  • Particle Size:
    90 µm
  • Max. pressure:
    5 bar
  • Capacity:
    5×1 ml
  • Description:
    HiTrap™ Capto™
  • Length:
    25 mm
  • Seq:
    01
  • pH range:
    2 - 12
  • Column hardware:
    PP
  • Pk:
    5

Specifications

About this item

HiTrap Capto IEX Selection Kit is designed for screening the optimal Capto ion exchange or Cytiva multimodal chromatography resin prior to scale-up.

  • Fast and easy selection. Five prepacked HiTrap columns, each containing a different IEX resin, including multimodal ion exchangers
  • Convenient way to screen for the optimal Cytiva IEX resin for a given application
  • Excellent for optimization studies, buffer composition, pH, flow rate, and other parameters can be optimized with small samples before scale-up
  • HiTrap column format is designed for use with a syringe, peristaltic pump, or chromatography system such as an ÄKTA system

This kit contains a variety of columns prepacked with anion and cation exchange resins, allowing users to screen for the most appropriate column prior to scale-up.

Delivery: Kit includes five 1 ml HiTrap™ columns prepacked with Capto™ Q (strong anion), Capto™ S (strong cation), Capto™ DEAE (weak anion), Capto™ MMC (multimodal cation), Capto™ adhere (multimodal strong anion), as well as connectors and instructions.