Order Entry
France
ContactUsLinkComponent
Cation exchange chromatography columns, HiTrap™ Capto™ SP ImpRes
Cation exchange chromatography columns, HiTrap™ Capto™ SP ImpRes
Supplier:  Cytiva
Certificates
undefined
Cation exchange chromatography columns, HiTrap™ Capto™ SP ImpRes
Supplier:  Cytiva
Specifications
  • Catalog No:
  • 17-5468-51
  • 17-5468-55
  • 29-4004-60
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • Storage:
  • 4...30 °C, 20% Ethanol, 0,2 M Sodium Acetate
  • 4...30 °C, 20% Ethanol, 0,2 M Sodium Acetate
  • 4...30 °C, 20% Ethanol, 0,2 M Sodium Acetate