Anion exchange chromatography columns, HiTrap™ Capto™ Q ImpRes
: Cytiva
HiTrap™ Capto™ Q ImpRes is a strong anion exchanger prepacked with BioProcess Capto™ ImpRes media for screening and small-scale protein purifications using ion exchange chromatography (IEX).
- High-throughput medium for intermediate and polishing protein purification, excellent choice for process-scale purification
- Rigid agarose matrix enables high bed heights and purification of viscous samples at high flow rates
- Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications
- Strong quartenary amine anion exchanger
The combined properties of high binding capacity, high flow rate and low back pressure reduces process cycle times and improves productivity at process scale.
- Catalog No:
- 17-5470-55
- 17-5470-51
- 29-4004-62
- Max. pressure:
- 5 bar
- 5 bar
- 5 bar
- Storage:
- 4...30 °C, 20% Ethanol
- 4...30 °C, 20% Ethanol
- 4...30 °C, 20% Ethanol
Frequently Bought Together