Surface protection papers
The surface paper protects laboratory benches against acids, toxic, corrosive and radioactive products and glass impact.
- White absorbent paper 100 g/m² with one-side PE coating 25 g/m²
- Available as sheets or on rolls
- Grade 604
For use with valuable materials.
Frequently Bought Together