Order Entry
France
ContactUsLinkComponent
Affinity chromatography columns, HiScreen™ Capto™ L
Affinity chromatography columns, HiScreen™ Capto™ L
Catalog # 17-5478-14
Supplier:  Cytiva
undefined
Affinity chromatography columns, HiScreen™ Capto™ L
Catalog # 17-5478-14
Supplier:  Cytiva

Specifications

  • Particle Size:
    85 µm
  • Max. pressure:
    3 bar
  • Capacity:
    1×4,7 ml
  • Description:
    HiScreen™ Capto™ L
  • Seq:
    01
  • pH range:
    2 - 10
  • Storage:
    2 to 8 °C, 20% Ethanol
  • Column hardware:
    PP
  • Bed dimensions:
    7,7×100 mm
  • Pk:
    1

Specifications

About this item

HiScreen™ Capto™ L columns are prepacked with Capto™ L, an affinity chromatography medium (resin) for capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs.

  • Prepacked, ready to use columns for convenient process development optimisation
  • Bed height of 10 cm allows method optimisation and parameter screening
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume (4,7 ml) gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity

The columns are an excellent choice for method optimisation and parameter screening.