Order Entry
France
ContactUsLinkComponent
 
Affinity chromatography media, MabSelect SuRe™
 :  Cytiva
 
undefined
Affinity chromatography media, MabSelect SuRe™
 :  Cytiva

 

  • pH range:
    3 - 12
  • Matrix:
    Highly cross-linked agarose, spherical
  • Ligand:
    Alkali-stabilized Protein A-derived (E. coli)
  • Storage temperature:
    2 - 8 °C
  • Binding capacity:
    ~35 mg hIgG/ml
  • Ligand coupling method:
    Epoxy activation

 

Frequently Bought Together