Order Entry
France
ContactUsLinkComponent
Hydrophobic interaction chromatography columns, HiTrap™ phenyl HP Sepharose™
Hydrophobic interaction chromatography columns, HiTrap™ phenyl HP Sepharose™
Supplier:  Cytiva
Certificates
undefined
Hydrophobic interaction chromatography columns, HiTrap™ phenyl HP Sepharose™
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 17-5195-01
  • 17-1351-01
  • Max. pressure:
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 13
  • 3 - 13
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Frequently Bought Together