Order Entry
France
ContactUsLinkComponent
Anti-IgG Sheep Antibody (HRP (Horseradish Peroxidase))
Anti-IgG Sheep Antibody (HRP (Horseradish Peroxidase))
Catalog # NA931-1ML
Supplier:  Cytiva
Anti-IgG Sheep Antibody (HRP (Horseradish Peroxidase))
Catalog # NA931-1ML
Supplier:  Cytiva

Specifications

  • Antibody type:
    Secondary
  • Antigen name:
    Immunoglobulin G
  • Antigen symbol:
    IgG
  • Conjugation:
    HRP (Horseradish Peroxidase)
  • ELISA:
    Yes
  • Host:
    Sheep
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Antibody fragment:
    Whole Ab
  • Storage buffer:
    Horseradish peroxidase conjugated antibody is supplied in phosphate buffered saline (sodium phosphate 0.1 M, NaCl 0.1 M) pH 7.5, containing 1% (w/v) bovine serum albumin and an anti-microbial agent
  • Storage temperature:
    2 - 8 °C
  • Immunogen:
    Mouse IgG
  • Tested applications:
    Western blotting
  • Pk:
    1 mL

Specifications

About this item

Highly species specific HRP-conjugated antibodies optimized for use with Amersham ECL Western Blotting Detection Reagents.

  • Optimized for use with our range of Amersham ECL Western blotting detection reagents
  • Soluble in cold water and hot water

The antibodies are supplied with dilution guidelines for relevant applications and should be stored at 2 °C to 8 °C.

Type:
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Mouse