Order Entry
France
Orders LinkContactUsLinkComponent
Inverted microscope, IX53
Inverted microscope, IX-53, Trinocular
  630-1872
Inverted microscope, IX53
  630-1872
Inverted microscope, IX53
Inverted microscope, IX53
Inverted microscope, IX-53, Trinocular
1 * 1 Pce
The IX53 is an evolutionary microscope, designed for applications on living cells, especially in research projects.
  • UIS2 generation optics with phase contrast
  • Dynamic structure and high definition imaging with minimal light loss
  • Possible modularity to terrain, fluorescence, polarization and interference contrast
Delivery information: Supplied with protective cover and two spare bulbs - Motorized models are also available on request.
  • Model:
    IX-53
  • Description:
    Inverted microscope
  • Type:
    Trinocular
  • Focusing system:
    Coaxial coarse and fine knobs
  • Cat. No.:
    630-1872
  • Observation method:
    Phase contrast
  • Condenser:
    N.A. 0,30
  • Nosepiece:
    Sextuple
  • Illumination:
    Halogen
Accessories
  •  
    Description
  • 630-0906
    IX MVR, attachable specimen guide (130×85 mm) with flexible right-hand vertical low drive control, for use with plain stage IX-SP
    1 * 1 Pce
  •  
    Description
  • 630-1874
    Camera adapter, U-TV1XC
    1 * 1 Pce - 548,00 €