Glucose determination tubes, VACUETTE®
Supplier: Greiner Bio-One
Certificates
About this item
PET, clear.
- Sterile
- With ridged safety twist cap or non-ridged pull cap
The VACUETTE® blood collection tubes made from practically unbreakable plastic have become well established on the market. They have made the drawing of blood a safer and more hygienic process. The use of PET plastic offers the advantage that the tubes are as clear as glass.
Frequently Bought Together