Order Entry
France
ContactUsLinkComponent
Hydrocortisone 97.0-102.0% (dried basis), micronised powder USP, Multi-Compendial, Macron Fine Chemicals™
Hydrocortisone 97.0-102.0% (dried basis), micronised powder USP, Multi-Compendial, Macron Fine Chemicals™
Certificates
undefined
Hydrocortisone 97.0-102.0% (dried basis), micronised powder USP, Multi-Compendial, Macron Fine Chemicals™
Certificates

Specification Test Results

  • Meets E.P. Chemical Specifications:
  • GMP Manufactured Product:
  • Meets U.S.P Requirements:
  • CAUTION: For Manufacturing, processing or repackaging:
  • Bulk Pharmaceutical Chemical:
  • USP - Assay (C₂₁H₃₀O₅) (dried basis):
    97.0 - 102.0 %
  • USP - Organic Impurities:
    Passes Test
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Loss on Drying:
    ≤ 1.0 %
  • USP - Residue on Ignition:
    ≤ 0.5 %
  • USP - Specific Rotation [Å]²⁵D (+):
    150 - 156 Degree
  • EP - Assay (C₂₁H₃₀O₅):
    97.0 - 103.0 %
  • EP - Identification A:
    Passes Test
  • EP - Identification B:
    Passes Test
  • EP - Specific Optical Rotation (+):
    162 - 168 Degree
  • EP - Related substances:impurity A:
    ≤ 0.2 %
  • EP - Related substances: impurity B:
    ≤ 0.2 %
  • EP - Related substances: impurity C:
    ≤ 0.5 %
  • EP - Related substances: impurity D:
    ≤ 0.5 %
  • EP - Related substances: impurity E:
    ≤ 0.5 %
  • EP - Related substances: impurity F:
    ≤ 0.3 %
  • EP - Related substances: impurity G:
    ≤ 0.4 %
  • EP - Related substances: impurity H:
    ≤ 0.15 %
  • EP - Related substances: impurity I:
    ≤ 0.5 %
  • EP - Related substances: impurity N:
    ≤ 0.15 %
  • EP - Related Substances: Unspecified impurities, Each:
    ≤ 0.10 %
  • EP - Related substances: Total impurities:
    ≤ 2.0 %
  • EP - Loss on Drying:
    ≤ 1.0 %

Specification Test Results

Related Information

Frequently Bought Together