Vert de bromocrésol ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-60-8
:
Formula:C₂₁H₁₄Br₄O₅S
MDL Number:MFCD00005874
:698.02 g/mol
Merck Index:14,1386
Storage Temperature:Température ambiante
EINECS:200-972-8
Related Information
Frequently Bought Together