D-(-)-Sorbitol
Catalog # PROORTC-PHR-1006
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Merck Index:13,08797
Boiling Pt:295 °C (1013 hPa)
Melting Pt:94…98 °C
Density:1.524 g/cm³ (20 °C)
Flash Pt:>100 °C
:182.17 g/mol
Storage Temperature:Ambient
EINECS:200-061-5
CAS Number:50-70-4
:98201-93-5
Formula:HOH₂C(CH(OH))₄CH₂OH
MDL Number:MFCD00004708