Potassium iodate 1/60 mol/l (0.1 N) in aqueous solution
Catalog # PRLSMC306814.1000
Supplier: PROLABO CMP
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:214 g/mol
Storage Temperature:Ambient
EINECS:231-831-9
CAS Number:7758-05-6
Formula:KlO₃
MDL Number:MFCD00011406