Order Entry
France
ContactUsLinkComponent
1-(Phenylsulphonyl)-2-indoleboronic acid
1-(Phenylsulphonyl)-2-indoleboronic acid
Catalog # MOLE30731776-1G
Supplier:  Molekula
CAS Number:  
1-(Phenylsulphonyl)-2-indoleboronic acid
Catalog # MOLE30731776-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
CAS Number:342404-46-0
Formula:C₁₄H₁₂BNO₄S
MDL Number:MFCD03086094