Order Entry
France
ContactUsLinkComponent
Anion exchange HPLC columns, biochemical analysis, NUCLEOGEL® SAX
Anion exchange HPLC columns, biochemical analysis, NUCLEOGEL® SAX
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Anion exchange HPLC columns, biochemical analysis, NUCLEOGEL® SAX
Supplier:  MACHEREY-NAGEL
Specifications
  • Catalog No:
  • MANA719469
  • MANA719600
  • Max. pressure:
  • 200 bar
  • 200 bar
  • pH range:
  • 1 - 13
  • 1 - 13
  • USP code:
  • L23
  • L23