Accessories for Memmert UN/UNplus and UF/UFplus Ovens
Supplier: MEMMERT
Certificates
About this item
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Propylene carbonate ≥99.0% (by GC), GPR RECTAPUR®
Supplier: VWR Chemicals
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
Accessories for desiccators, Nalgene®
Supplier: Thermo Fisher Scientific
VWR®, Single-Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
Accessories for MA series
Supplier: Sartorius Balances
Accessories
Supplier: Merck
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
PUMPHEAD VAMPIR SAMPLER 1 * 1 items
Supplier: BURKLE