Order Entry
France
ContactUsLinkComponent
HPLC columns, NUCLEOSIL® C2
HPLC columns, NUCLEOSIL® C2
Supplier:  MACHEREY-NAGEL
Certificates
undefined
HPLC columns, NUCLEOSIL® C2
Supplier:  MACHEREY-NAGEL
Specifications
  • Catalog No:
  • MANA721030.30
  • MANA720089.46
  • Carbon load:
  • 3,5%
  • 3,5%
  • Endcapped:
  • No, No
  • No, No
  • pH range:
  • 2 - 8
  • 2 - 8
  • USP code:
  • L16, L16
  • L16, L16