SPE columns, CHROMABOND® SA
Supplier: MACHEREY-NAGEL
Certificates
About this item
CHROMABOND® SA is a benzenesulphonic acid modified silica cation exchanger (SCX) for SPE.
- Base material: Silica
- Strongly acidic cation exchanger
- Additionally hydrophobic and π-π interactions (benzene ring)
Recommended for ion exchange of organic compounds (e.g. amines, amino acids, triazines) from aqueous matrix.
Cation exchange capacity: ~0,5 meq/g
Specifications
- Catalog No:
- MANA738141.100M
- MANA730425
- MANA731832
- MANA731831
- MANA731833
- MANA730076
- MANA730275
- MANA730077
- MANA730212
- MANA730077.250
- pH range:
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- 2 - 8
- Surface area:
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g
- 500 m²/g