Order Entry
France
ContactUsLinkComponent
SPE columns, CHROMABOND® C18 ec f (fast flow)
SPE columns, CHROMABOND® C18 ec f (fast flow)
Supplier:  MACHEREY-NAGEL
Certificates
undefined
SPE columns, CHROMABOND® C18 ec f (fast flow)
Supplier:  MACHEREY-NAGEL
Specifications
  • Catalog No:
  • MANA730010
  • MANA730016
  • MANA730269
  • Carbon load:
  • 14%
  • 14%
  • 14%
  • Endcapped:
  • Yes
  • Yes
  • Yes
  • pH range:
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • Surface area:
  • 500 m²/g
  • 500 m²/g
  • 500 m²/g