Order Entry
France
Orders LinkContactUsLinkComponent
HPLC columns, NUCLEOSIL® C6H5
HPLC columns, NUCLEOSIL® C6H5
Supplier:   MACHEREY-NAGEL
HPLC columns, NUCLEOSIL® C6H5
Supplier:  MACHEREY-NAGEL
Certificates
HPLC columns, NUCLEOSIL® C6H5
4 Options Available Below
Product Details & Documents
NUCLEOSIL® C6H5 is a non-endcapped phenyl modified silica phase for HPLC.
  • Robust spherical silica gel
  • Weakly hydrophobic, non-endcapped phase with aromatic selectivity for RP chromatography
  • Separation mechanism is especially based on π–π interactions as well as on hydrophobic and some residual silanol interactions
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Specifications for Product Family
Specifications
  • Catalog No:
  • MANA720956.46
  • MANA720019.40
  • MANA720019.46
  • MANA721137.30
  • Carbon load:
  • 8%
  • 8%
  • 8%
  • 8%
  • Endcapped:
  • No|No
  • No|No
  • No|No
  • No|No
  • pH range:
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • 2 - 8
  • USP code:
  • L11|L11
  • L11|L11
  • L11|L11
  • L11|L11
Specifications
Product Family Options
Analytical columns
Product Information
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Analytical
    C6H5
    100 Å
    5 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 724,00 €
    724,00 €
    1 * 1 Pce
       724,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA720956.46
    Carbon load
    8%
    Endcapped
    No|No
    pH range
    2 - 8
    USP code
    L11|L11
  • Analytical
    C6H5
    100 Å
    7 µm
    4,00 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 686,00 €
    686,00 €
    1 * 1 Pce
       686,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA720019.40
    Carbon load
    8%
    Endcapped
    No|No
    pH range
    2 - 8
    USP code
    L11|L11
  • Analytical
    C6H5
    100 Å
    7 µm
    4,60 mm
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 724,00 €
    724,00 €
    1 * 1 Pce
       724,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA720019.46
    Carbon load
    8%
    Endcapped
    No|No
    pH range
    2 - 8
    USP code
    L11|L11
Guard columns
Product Information
  • Column Format
    Phase
    Pore size
    Particle Size
    Ø int.
    Length
    Availability
    Price
  • Guard column
    C6H5
    100 Å
    5 µm
    3,00 mm
    4 mm
    Log in for availability
    Log in for availability
    1 * 3 Pce
    1 * 3 Pce - 234,00 €
    234,00 €
    1 * 3 Pce
       234,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA721137.30
    Carbon load
    8%
    Endcapped
    No|No
    pH range
    2 - 8
    USP code
    L11|L11