Order Entry
France
ContactUsLinkComponent
Reversed phase HPLC columns, biochemical analysis, NUCLEOSIL® PPN
Reversed phase HPLC columns, biochemical analysis, NUCLEOSIL® PPN
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Reversed phase HPLC columns, biochemical analysis, NUCLEOSIL® PPN
Supplier:  MACHEREY-NAGEL
Specifications
  • Catalog No:
  • MANA720258.40
  • MANA720252.40
  • Max. pressure:
  • 250 bar
  • 250 bar
  • pH range:
  • 1 – 9
  • 1 – 9
  • USP code:
  • L1
  • L1