Uvex pheos CX2 safety spectacles demonstrate excellent performance
Suitable for most disciplines.
Winner at work!
The new uvex pheos CX2, takes uvex’s best selling spectacle to new heights. The X-tended eye shield and brow guard, with injection moulded, soft component technology, provides reliable protection against splash and particle ingress. The four-point frame check ensures the spectacle fits the wearer perfectly to deliver reliable protection. The distinctive design and shape of the side arms, together with the innovative X-Twist technology, allows the uvex pheos CX2 to adapt to individual head widths providing a comfortable, non slip fit. uvex pheos CX2 safety spectacles achieve outstanding performance for the protection in daily working life.
- X-Twist technology
 - Scratch resistant and anti-fog
 - X-tended eye shield
 - Fold easily
 - Perfect ventilation
Log in to see your contract pricing and availability
| Anti-UV (5-2,5), lenses | Colour | Cat. No. | 
|---|---|---|
| Grey PC | White/black | 111-2148E | 
| Clear PC | Black/blue | 111-2149E | 
| Clear PC | Black/grey | 111-2150E | 
| Clear PC | Red/grey | 111-2151E | 
| Clear PC | Blue/grey | 111-2332E | 