Orders LinkContactUsLinkComponent
372075Featuregifimg.gif
Microflex® 93-260 nitrile and neoprene double-coated disposable gloves
Thinnest chemical resistant synthetic composite disposable glove.
Microflex 93-260 nitrile and neoprene double-coated disposable gloves | Avantor
  • Three layer design for superior protection against harsh chemicals including acids, solvents and bases
  • Thin mil construction provides enhanced tactility and dexterity
  • Extra soft material and ergonomic design for outstanding fit, feel and flexibility for longer wear time
  • Lower acceptable pin hole rate (0,65 AQL) and extended cuff for reliable protection against hazardous substances
  • Silicone-free formulation and processing ensure better product protection