Order Entry
Finland
ContactUsLinkComponent
EMITTER TAPERED TIP, DISTAL COATED 360UM OD X 20UM ID X 50MM, 13 DEG, PK5 1 * 5 items
Catalog # SGEA063240100
New Product
EMITTER TAPERED TIP, DISTAL COATED 360UM OD X 20UM ID X 50MM, 13 DEG, PK5 1 * 5 items
Catalog # SGEA063240100
New Product