Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
6ft Nexus Horizontal Clean Bench. 21inch deep work area with stainless steel interior. Includes UV and unassembled base stand. 230V SC 1 * 1 items
Catalog # LBCP5360830
Supplier:
LABCONCO
New Product
Expand image
6ft Nexus Horizontal Clean Bench. 21inch deep work area with stainless steel interior. Includes UV and unassembled base stand. 230V SC 1 * 1 items
Catalog # LBCP5360830
Supplier:
LABCONCO
New Product