Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Self Seal Closure, With Steam Indicator Dot, Size: 14 x 24 1 * 500 items
Catalog # KACC01424TWBBSSSD
Supplier:
KEYSTONE CLEANROOM PRODUCTS
New Product
Expand image
Autoclavable Tyvek Breather Bag, With Easy Chevron Peel Feature & Self Seal Closure, With Steam Indicator Dot, Size: 14 x 24 1 * 500 items
Catalog # KACC01424TWBBSSSD
Supplier:
KEYSTONE CLEANROOM PRODUCTS
New Product