Order Entry
Finland
ContactUsLinkComponent
Human pan IFN alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, recombinant human IFN alpha ELISA standard, and standard reconstitution buffer. This mat 1 * 1.920 Tests
Catalog # ANTIA270501-2096
Supplier:  Antibodies.com
New Product
Human pan IFN alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-HRP, recombinant human IFN alpha ELISA standard, and standard reconstitution buffer. This mat 1 * 1.920 Tests
Catalog # ANTIA270501-2096
Supplier:  Antibodies.com