Order Entry
Finland
ContactUsLinkComponent
Mouse pan IFN alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, recombinant mouse IFN alpha ELISA standard, and standard reconstitution buffer. This matc 1 * 576 Tests
Catalog # ANTIA270447-696
Supplier:  Antibodies.com
New Product
Mouse pan IFN alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody, Streptavidin-AP, recombinant mouse IFN alpha ELISA standard, and standard reconstitution buffer. This matc 1 * 576 Tests
Catalog # ANTIA270447-696
Supplier:  Antibodies.com