Order Entry
Finland
ContactUsLinkComponent
Mouse IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant mouse IL-10 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 1.920 Tests
Catalog # ANTIA270520-2096
Supplier:  Antibodies.com
New Product
Mouse IL-10 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant mouse IL-10 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 1.920 Tests
Catalog # ANTIA270520-2096
Supplier:  Antibodies.com