Order Entry
Finland
ContactUsLinkComponent
Human IL-13 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant human IL-13 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
Catalog # ANTIA270403-696
Supplier:  Antibodies.com
New Product
Human IL-13 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant human IL-13 ELISA standard, and standard reconstitution buffer. This matched antibody 1 * 576 Tests
Catalog # ANTIA270403-696
Supplier:  Antibodies.com