Order Entry
Finland
ContactUsLinkComponent
Human IL-17A/F Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-AP, recombinant human IL-17A/F ELISA standard, and standard reconstitution buffer. This matched an 1 * 1.920 Tests
Catalog # ANTIA325367-2096
Supplier:  Antibodies.com
New Product
Human IL-17A/F Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection anti body, Streptavidin-AP, recombinant human IL-17A/F ELISA standard, and standard reconstitution buffer. This matched an 1 * 1.920 Tests
Catalog # ANTIA325367-2096
Supplier:  Antibodies.com