Mouse IgG Matched Antibody Pair Kit includes an unlabeled polyclonal capture antibody, an HRP-labelled polyclonal detection antibody , native mouse IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can be used to 1 * 576 Tests
Catalog # ANTIA325382-696
Supplier: Antibodies.com