Order Entry
Finland
ContactUsLinkComponent
Mouse SFXN1/TCC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse SFXN1/TCC in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5673-48
Supplier:  ANTIBODIES.COM
New Product
Mouse SFXN1/TCC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse SFXN1/TCC in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA5673-48
Supplier:  ANTIBODIES.COM