Rat NOR1/TEC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat NOR1/TEC in tissue homogenates or other biological fluids. 1 * 96 Tests
Catalog # ANTIA5650-96
Supplier: Antibodies.com