Order Entry
Finland
ContactUsLinkComponent
Rat CXCL17/DMC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat CXCL17/DMC in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6824-48
Supplier:  Antibodies.com
New Product
Rat CXCL17/DMC ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat CXCL17/DMC in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA6824-48
Supplier:  Antibodies.com