Order Entry
Finland
Orders LinkContactUsLinkComponent
 

2-Amino-4,5-dichlorophenol

Bottle
Catalog # APOSOR16946-25G
Irritant
New Product
Supplier:   Apollo Scientific
2-Amino-4,5-dichlorophenol
Catalog # APOSOR16946-25G
Supplier:  Apollo Scientific
New Product
CAS Number:  28443-57-4
2-Amino-4,5-dichlorophenol

Bottle

CAS Number:  28443-57-4
Price
1 010,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  4,5-Dichloro-2-hydroxyaniline
  • CAS Number:28443-57-4
  • Formula:C₆H₅Cl₂NO
  • MDL Number:MFCD12406838
  • Molecular Weight:178.016 g/mol
  • Melting Pt:170…175 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications