Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
COLUMBIA CNA AGAR (SHEEP BLOOD 5%) 90 MM 1 * 20 items
Catalog # LIOF11024
Supplier:
LIOFILCHEM
New Product
Certificates
Expand image
COLUMBIA CNA AGAR (SHEEP BLOOD 5%) 90 MM 1 * 20 items
Catalog # LIOF11024
Supplier:
LIOFILCHEM
New Product
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation