Order Entry
Finland
ContactUsLinkComponent
EXAMINATION AND DISPOSABLE PROTECTIVE GLOVE, MADE OF NITRILE BUTADIENE RUBBER (NBR), GREEN, POWDER-FREE, ACCELERATOR-FREE, NON-STERILE, AMBIDEXTROUS, FOR SINGLE USE. COMPLIANT WITH MD REGULATION (EU) 2017/745 AND PPE REGULATION (EU) 2016/425. 1 * 200 items
Catalog # VWRI112-6647
Supplier:  VWR Collection
New Product
EXAMINATION AND DISPOSABLE PROTECTIVE GLOVE, MADE OF NITRILE BUTADIENE RUBBER (NBR), GREEN, POWDER-FREE, ACCELERATOR-FREE, NON-STERILE, AMBIDEXTROUS, FOR SINGLE USE. COMPLIANT WITH MD REGULATION (EU) 2017/745 AND PPE REGULATION (EU) 2016/425. 1 * 200 items
Catalog # VWRI112-6647
Supplier:  VWR Collection
New Product