VWR® Xtra Nitrile Green Glove
Catalog # VWRI112-6649
Supplier: VWR Collection
New Product
Specifications
- Glove size:M
- Ambidextrous / hand-specific:Ambidextrous
- Disposable / reusable:Disposable
- Latex / latex-free:Latex-free
- Powdered / powder-free:Powder-free
- Colour:Green
- Length:240 mm
- Seq:3
- Thickness:0,6 mil
Specifications
About this item
These single-use, disposable examination protective gloves are made from nitrile and are developed with sustainability in mind. They are suitable for use in medical applications, food processing, and for general lab requirements in non-hazardous areas.
Non sterile, powder-free
- Free from latex, chlorine, and accelarators
- AQL 1.0
- Food contact approved
The environmental impact of this glove has been assessed via Life Cycle Analysis meeting ISO 14040 and ISO 14044. Reduce your eco-footprint while still wearing a glove that is skin-friendly, and provides superior barrier protection against blood-borne pathogens and viruses.
Certifications:
EN ISO 374-1 Type B (JKT), EN 374-5 VIRUS, MDR Class I; PPE Cat. III; AQL 1.5, food contact approved.
Packaging:
Supplied 200 per bag (or 180 for size XL). 10 dispensers/carton. Less packaging waste.