Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Masterflex Transfer Tubing, Flexible PVC, non-DEHP, 3/16 x 5/16, 50 ft 1 * 50 Feet
Catalog # MFLX96505-08
Supplier:
Avantor Fluid Handling
New Product
Expand image
Masterflex Transfer Tubing, Flexible PVC, non-DEHP, 3/16 x 5/16, 50 ft 1 * 50 Feet
Catalog # MFLX96505-08
Supplier:
Avantor Fluid Handling
New Product