Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FREEZONE 6L -50C CONSOLE FREEZE DRYER, PTFE-COATED COLLECTOR, STOPPERING TRAY DRYER, 230V, NATURAL REFRIGERATION GAS 1 * 1 items
Catalog # LBCP700626090
Supplier:
LABCONCO
New Product
Expand image
FREEZONE 6L -50C CONSOLE FREEZE DRYER, PTFE-COATED COLLECTOR, STOPPERING TRAY DRYER, 230V, NATURAL REFRIGERATION GAS 1 * 1 items
Catalog # LBCP700626090
Supplier:
LABCONCO
New Product