Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TISSUE SECTION BATH 1 * 1 items
Catalog # SHANA84600060
Supplier:
Epredia
New Product
Expand image
TISSUE SECTION BATH 1 * 1 items
Catalog # SHANA84600060
Supplier:
Epredia
New Product