Order Entry
Finland
ContactUsLinkComponent
Masterflex Transfer Tubing, Poly Flex, Red, 1/4 x 3/8 , 100 ft/30,48M 1 * 3.048 cm
Catalog # MFLX92419-15
New Product
Masterflex Transfer Tubing, Poly Flex, Red, 1/4 x 3/8 , 100 ft/30,48M 1 * 3.048 cm
Catalog # MFLX92419-15
New Product